missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals L(-)-Glucose, 98%
$79.69 - $633.27
Chemical Identifiers
CAS | 921-60-8 |
---|---|
Molecular Formula | C6H12O6 |
Molecular Weight (g/mol) | 180.16 |
MDL Number | MFCD00148913 |
InChI Key | GZCGUPFRVQAUEE-VANKVMQKSA-N |
Synonym | l-glucose, l---glucose, levoglucose, l--glucose, 2s,3r,4s,5s-2,3,4,5,6-pentahydroxyhexanal, aldehydo-l-glucose, aldehydo-l-gluco-hexose, d-glucose-13c6,d7, levoglucose usan, glucose, l-isomer |
PubChem CID | 10954115 |
ChEBI | CHEBI:37626 |
IUPAC Name | (2S,3R,4S,5S)-2,3,4,5,6-pentahydroxyhexanal |
SMILES | C(C(C(C(C(C=O)O)O)O)O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC241922500
|
Thermo Scientific Chemicals
241922500 |
250 mg | Glass bottle |
Each for $79.69
|
|
||||
AC241920010
|
Thermo Scientific Chemicals
241920010 |
1 g | Glass bottle |
Each for $199.80
|
|
||||
AC241920050
|
Thermo Scientific Chemicals
241920050 |
5 g | Glass bottle |
Each for $633.27
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
921-60-8 | |
180.16 | |
GZCGUPFRVQAUEE-VANKVMQKSA-N | |
10954115 | |
(2S,3R,4S,5S)-2,3,4,5,6-pentahydroxyhexanal |
C6H12O6 | |
MFCD00148913 | |
l-glucose, l---glucose, levoglucose, l--glucose, 2s,3r,4s,5s-2,3,4,5,6-pentahydroxyhexanal, aldehydo-l-glucose, aldehydo-l-gluco-hexose, d-glucose-13c6,d7, levoglucose usan, glucose, l-isomer | |
CHEBI:37626 | |
C(C(C(C(C(C=O)O)O)O)O)O |
Specifications
921-60-8 | |
White | |
97.5% min. (ELSD) (HPLC) | |
C6H12O6 | |
250 mg | |
− 52.00 | |
(10% in water) Clear colorless | |
C(C(C(C(C(C=O)O)O)O)O)O | |
180.16 | |
CHEBI:37626 | |
98% | |
L(-)-Glucose |
153.0°C to 156.0°C | |
Authentic | |
Glass bottle | |
MFCD00148913 | |
− 52.00 (20.00°C c=10,H2O,NH3) | |
l-glucose, l---glucose, levoglucose, l--glucose, 2s,3r,4s,5s-2,3,4,5,6-pentahydroxyhexanal, aldehydo-l-glucose, aldehydo-l-gluco-hexose, d-glucose-13c6,d7, levoglucose usan, glucose, l-isomer | |
GZCGUPFRVQAUEE-VANKVMQKSA-N | |
(2S,3R,4S,5S)-2,3,4,5,6-pentahydroxyhexanal | |
10954115 | |
180.16 | |
Crystalline Powder |
Safety and Handling
EINECSNumber : 213-068-3
RUO – Research Use Only