Learn More
L-Glutamic acid 1-methyl ester, 98%
CAS: 6384-08-3 | C8H12N2 | 136.20 g/mol
$71.49 - $268.33
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6327903
|
Thermo Scientific Chemicals
H6327903 |
1 g |
Each for $71.49
|
|
|||||
|
AAH6327906
|
Thermo Scientific Chemicals
H6327906 |
5 g |
Each for $268.33
|
|
|||||
Description
L-Glutamic acid 1-methyl ester nonessential amino acid that is important to the mammalian central nervous system. It is also a neurotransmitter for cone photoreceptors in the human brain and is used as a treatment for patients who have liver disease accompanied by encephalopathy (a condition termed âhepatic comaâ).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in water.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Chemical Identifiers
| 6384-08-3 | |
| MFCD00171677 |
Specifications
| 145°C (decomposition) | |
| White | |
| MFCD00171677 | |
| 2,5-dimethyl-1,4-benzenediamine, 4-amino-2,5-dimethylaniline, 2,5-dimethyl-1,4-phenylenediamine, 2,5-dimethyl-p-phenylenediamine, 2,5-diamino-p-xylene, 2,5-dimethylphenylene-1,4-diamine, 2,5-dimethyl-1,4-benzoquinonediamine, ccris 8118, 1,4-dimethylphenylene-2,5-diamine, unii-re400vn2b4 | |
| BWAPJIHJXDYDPW-UHFFFAOYSA-N | |
| 2,5-dimethylbenzene-1,4-diamine | |
| 136.20 | |
| CHEBI:53613 | |
| 98% | |
| L-Glutamic acid 1-methyl ester |
| 6384-08-3 | |
| C8H12N2 | |
| 1707273 | |
| Soluble in water. | |
| CC1=CC(=C(C=C1N)C)N | |
| 1 g | |
| 160824 | |
| 161.16 | |
| Powder |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only