missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Glutamic Acid, ≥99%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0210179305

 View more versions of this product

Catalog No. ICN10179305


Explore available promotions

Description

Description

Molecular biology, Cell Biology
Specifications

Specifications

L-Glutamic Acid
White
C5H9NO4
l-glutamic acid, glutamic acid, 2s-2-aminopentanedioic acid, h-glu-oh, s-2-aminopentanedioic acid, glutaminol, l-glutaminic acid, glutacid, glutamicol, glutamidex
NC(CCC(O)=O)C(O)=O
(2S)-2-aminopentanedioic acid
147.13
CHEBI:16015
Powder
56-86-0
≥99%
MFCD00002634
WHUUTDBJXJRKMK-UHFFFAOYNA-N
1.538g/mL at 20°C (Lit.)
5 kg
33032
147.1

Chemical Identifiers

56-86-0
147.13
WHUUTDBJXJRKMK-UHFFFAOYNA-N
33032
NC(CCC(O)=O)C(O)=O
C5H9NO4
MFCD00002634
l-glutamic acid, glutamic acid, 2s-2-aminopentanedioic acid, h-glu-oh, s-2-aminopentanedioic acid, glutaminol, l-glutaminic acid, glutacid, glutamicol, glutamidex
CHEBI:16015
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-293-7

Recommended Storage : Store at room temperature (15° to 30°C).

SDS
Documents

Documents

Promotions

Promotions