missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Glutamic Acid, ≥99%, MP Biomedicals™
Supplier: MP Biomedicals Inc 021017931
Description
Molecular biology, Cell BiologySpecifications
L-Glutamic Acid | |
White | |
C5H9NO4 | |
l-glutamic acid, glutamic acid, 2s-2-aminopentanedioic acid, h-glu-oh, s-2-aminopentanedioic acid, glutaminol, l-glutaminic acid, glutacid, glutamicol, glutamidex | |
NC(CCC(O)=O)C(O)=O | |
2-aminopentanedioic acid | |
147.13 | |
CHEBI:16015 | |
Powder |
56-86-0 | |
≥99% | |
MFCD00002634 | |
WHUUTDBJXJRKMK-UHFFFAOYNA-N | |
1.538g/mL at 20°C (Lit.) | |
100 g | |
33032 | |
147.1 |
Chemical Identifiers
56-86-0 | |
147.13 | |
WHUUTDBJXJRKMK-UHFFFAOYNA-N | |
33032 | |
NC(CCC(O)=O)C(O)=O |
C5H9NO4 | |
MFCD00002634 | |
l-glutamic acid, glutamic acid, 2s-2-aminopentanedioic acid, h-glu-oh, s-2-aminopentanedioic acid, glutaminol, l-glutaminic acid, glutacid, glutamicol, glutamidex | |
CHEBI:16015 |
Safety and Handling
EINECSNumber : 200-293-7
Recommended Storage : Store at room temperature (15° to 30°C).