missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Glutamic Acid, free acid, ≥99%, MP Biomedicals™
$63.65 - $136.61
Chemical Identifiers
CAS | 142-47-2 |
---|---|
Molecular Formula | C5H8NNaO4 |
Molecular Weight (g/mol) | 169.11 |
MDL Number | MFCD00150138 |
InChI Key | LPUQAYUQRXPFSQ-UHFFFAOYNA-M |
Synonym | natriumglutaminat, sodium glutamate, glutamate sodium, monosodioglutammato, sodium l-glutamate, glutammato monosodico, monosodium l-glutamate, sodium hydrogen glutamate, monosodium glutamate, glutamic acid, monosodium salt |
PubChem CID | 86748263 |
IUPAC Name | sodium 2-amino-4-carboxybutanoate |
SMILES | [Na+].NC(CCC(O)=O)C([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN10180080
|
MP Biomedicals Inc
0210180080 |
100 g |
Each for $63.65
|
|
|||||
ICN10180091
|
MP Biomedicals Inc
0210180091 |
1 kg |
Each for $136.61
|
|
|||||
Chemical Identifiers
142-47-2 | |
169.11 | |
LPUQAYUQRXPFSQ-UHFFFAOYNA-M | |
86748263 | |
[Na+].NC(CCC(O)=O)C([O-])=O |
C5H8NNaO4 | |
MFCD00150138 | |
natriumglutaminat, sodium glutamate, glutamate sodium, monosodioglutammato, sodium l-glutamate, glutammato monosodico, monosodium l-glutamate, sodium hydrogen glutamate, monosodium glutamate, glutamic acid, monosodium salt | |
sodium 2-amino-4-carboxybutanoate |
Specifications
142-47-2 | |
≥99% | |
MFCD00150138 | |
LPUQAYUQRXPFSQ-UHFFFAOYNA-M | |
1.538g/mL at 20°C | |
100 g | |
86748263 | |
Powder |
White | |
C5H8NNaO4 | |
natriumglutaminat, sodium glutamate, glutamate sodium, monosodioglutammato, sodium l-glutamate, glutammato monosodico, monosodium l-glutamate, sodium hydrogen glutamate, monosodium glutamate, glutamic acid, monosodium salt | |
[Na+].NC(CCC(O)=O)C([O-])=O | |
sodium 2-amino-4-carboxybutanoate | |
169.11 | |
187.13 | |
L-Glutamic acid Monosodium Salt |
Safety and Handling
EINECSNumber : 205-538-1
Recommended Storage : Store at room temperature (15 to 30°C).