Learn More
L-Glutamic acid monopotassium salt monohydrate, 97+%
CAS: 6382-01-0 | C5H11KNO5 | 204.243 g/mol
$124.62 - $430.74
Chemical Identifiers
| CAS | 6382-01-0 |
|---|---|
| Molecular Formula | C5H11KNO5 |
| Molecular Weight (g/mol) | 204.243 |
| MDL Number | MFCD00150343 |
| InChI Key | VMWJTBYKMYBHOC-QTNFYWBSSA-N |
| Synonym | l-glutamic acid hydrate potassium, l-glutamicacid,monopotassiumsalt,monohydrate |
| PubChem CID | 87237506 |
| IUPAC Name | (2S)-2-aminopentanedioic acid;potassium;hydrate |
| SMILES | C(CC(=O)O)C(C(=O)O)N.O.[K] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1723230
|
Thermo Scientific Chemicals
A1723230 |
250 g |
Each for $124.62
|
|
|||||
|
AAA172320B
|
Thermo Scientific Chemicals
A172320B |
1000 g |
Each for $430.74
|
|
|||||
Description
L-Glutamic acid monopotassium salt monohydrate acts as a common excitatory neurotransmitter like the agonist at kainate, N-methyl-D-aspartate receptor (NMDA) and quisqualate receptors in central nervous system. It is also used in cell culture as a component of MEM non-essential amino acids solution..
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsL-Glutamic acid monopotassium salt monohydrate acts as a common excitatory neurotransmitter like the agonist at kainate, N-methyl-D-aspartate receptor (NMDA) and quisqualate receptors in central nervous system. It is also used in cell culture as a component of MEM non-essential amino acids solution..
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
| 6382-01-0 | |
| 204.243 | |
| VMWJTBYKMYBHOC-QTNFYWBSSA-N | |
| 87237506 | |
| C(CC(=O)O)C(C(=O)O)N.O.[K] |
| C5H11KNO5 | |
| MFCD00150343 | |
| l-glutamic acid hydrate potassium, l-glutamicacid,monopotassiumsalt,monohydrate | |
| (2S)-2-aminopentanedioic acid;potassium;hydrate |
Specifications
| 115°C to 118°C | |
| C5H11KNO5 | |
| l-glutamic acid hydrate potassium, l-glutamicacid,monopotassiumsalt,monohydrate | |
| VMWJTBYKMYBHOC-QTNFYWBSSA-N | |
| (2S)-2-aminopentanedioic acid;potassium;hydrate | |
| 204.243 | |
| 203.25 (185.23 Anhydrous) | |
| L-Glutamic acid monopotassium salt monohydrate |
| 6382-01-0 | |
| MFCD00150343 | |
| Soluble in water. | |
| C(CC(=O)O)C(C(=O)O)N.O.[K] | |
| 250 g | |
| 87237506 | |
| ≥97% |
Safety and Handling
RTECSNumber : MA1450000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only