missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

γ-L-Glutamyl-p-nitroanilide, Hydrochloride, Fisher BioReagents

$1185.63 - $1185.63

Chemical Identifiers

CAS 67953-08-6
Molecular Formula C11H14ClN3O5
Molecular Weight (g/mol) 303.70
MDL Number MFCD00039054
InChI Key WMRCHNNHOJNRSL-UHFFFAOYNA-N
Synonym gamma-l-glutamyl-p-nitroanilide, n-4-nitrophenyl-l-glutamine monohydrochloride, l-2-amino-4'-nitroglutaranilic acid hydrochloride, 2-amino-4-4-nitrophenyl carbamoyl butanoic acid hydrochloride, l-, a-glutamyl-p-nitroanilide, l-glutamic acid, a-p-nitroanilide hydrochloride, l-glutamic acid gamma-p-nitroanilide hydrochlori, l-glutamic acid gamma-p-nitroanilide hydrochloride, gamma-glutamyl transpeptidase substrate
IUPAC Name N-(3-carbamoyl-1-carboxypropyl)-4-nitroanilinium chloride
SMILES [Cl-].NC(=O)CCC([NH2+]C1=CC=C(C=C1)[N+]([O-])=O)C(O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BP26565
View Documents
Fisher BioReagents
BP26565
5 g Amber Glass
Each for $1,185.63
Case of 9 Each for $10,670.67
 
Description

Description

γ-L-Glutamyl-p-Nitroanilide, Hydrochloride is a water soluble form of GPNA suitable for use as substrate for γ-glutamyl transpeptidase.
Specifications

Chemical Identifiers

67953-08-6
303.70
WMRCHNNHOJNRSL-UHFFFAOYNA-N
N-(3-carbamoyl-1-carboxypropyl)-4-nitroanilinium chloride
C11H14ClN3O5
MFCD00039054
gamma-l-glutamyl-p-nitroanilide, n-4-nitrophenyl-l-glutamine monohydrochloride, l-2-amino-4'-nitroglutaranilic acid hydrochloride, 2-amino-4-4-nitrophenyl carbamoyl butanoic acid hydrochloride, l-, a-glutamyl-p-nitroanilide, l-glutamic acid, a-p-nitroanilide hydrochloride, l-glutamic acid gamma-p-nitroanilide hydrochlori, l-glutamic acid gamma-p-nitroanilide hydrochloride, gamma-glutamyl transpeptidase substrate
[Cl-].NC(=O)CCC([NH2+]C1=CC=C(C=C1)[N+]([O-])=O)C(O)=O

Specifications

67953-08-6
Amber Glass
MFCD00039054
gamma-l-glutamyl-p-nitroanilide, n-4-nitrophenyl-l-glutamine monohydrochloride, l-2-amino-4'-nitroglutaranilic acid hydrochloride, 2-amino-4-4-nitrophenyl carbamoyl butanoic acid hydrochloride, l-, a-glutamyl-p-nitroanilide, l-glutamic acid, a-p-nitroanilide hydrochloride, l-glutamic acid gamma-p-nitroanilide hydrochlori, l-glutamic acid gamma-p-nitroanilide hydrochloride, gamma-glutamyl transpeptidase substrate
[Cl-].NC(=O)CCC([NH2+]C1=CC=C(C=C1)[N+]([O-])=O)C(O)=O
303.70
Powder or Solid
Beige
C11H14ClN3O5
5 g
WMRCHNNHOJNRSL-UHFFFAOYNA-N
N-(3-carbamoyl-1-carboxypropyl)-4-nitroanilinium chloride
303.7
γ-L-Glutamyl-p-Nitroanilide, Hydrochloride
Videos
Safety and Handling

Safety and Handling

Recommended Storage : 0° to 5°C

SDS
missing translation for 'documents'

missing translation for 'documents'