missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Histidine hydrochloride monohydrate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 411731000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
L-Histidine hydrochloride monohydrate | |
200ppm max. | |
3 to 5 (2.5% soln. at 25°C) | |
10ppm max. | |
C6H12ClN3O3 | |
MFCD00151027 | |
25, 513 | |
10, 4617 | |
Plastic Bottle | |
CMXXUDSWGMGYLZ-XRIGFGBMSA-N | |
(2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid;hydrate;hydrochloride | |
209.63 | |
300ppm max. | |
98% | |
+ 9.90 (20.00°C c=11,6 N HCl) | |
Crystalline Powder or Crystals |
98% | |
5934-29-2 | |
White | |
98% | |
Authentic | |
10ppm max. | |
7% to 10% (3 hrs) (145 to 150°C) | |
0.5% max. (TLC) | |
l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.h2o, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrate | |
C1=C(NC=N1)CC(C(=O)O)N.O.Cl | |
+ 9.90 | |
165377 | |
209.64 | |
0.1% max. | |
100 g |
Chemical Identifiers
5934-29-2 | |
209.63 | |
CMXXUDSWGMGYLZ-XRIGFGBMSA-N | |
165377 | |
C1=C(NC=N1)CC(C(=O)O)N.O.Cl |
C6H12ClN3O3 | |
MFCD00151027 | |
l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.h2o, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrate | |
(2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid;hydrate;hydrochloride |
Safety and Handling
RTECSNumber : MS3119000
RUO – Research Use Only