missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Histidine Monohydrochloride Monohydrate, MP Biomedical
$48.36 - $997.93
Chemical Identifiers
CAS | 5934-29-2 |
---|---|
Molecular Formula | C6H12ClN3O3 |
Molecular Weight (g/mol) | 209.63 |
MDL Number | MFCD00151027 |
InChI Key | CMXXUDSWGMGYLZ-XRIGFGBMSA-N |
Synonym | l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.h2o, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrate |
PubChem CID | 165377 |
IUPAC Name | (2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid;hydrate;hydrochloride |
SMILES | C1=C(NC=N1)CC(C(=O)O)N.O.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN10195725
|
MP Biomedicals Inc
0210195725 |
25 g |
Each for $48.36
|
|
|||||
ICN10195780
|
MP Biomedicals Inc
0210195780 |
100 g |
Each for $145.09
|
|
|||||
ICN10195790
|
MP Biomedicals Inc
0210195790 |
500 g |
Each for $635.55
|
|
|||||
ICN10195791
|
MP Biomedicals Inc
0210195791 |
1 kg |
Each for $997.93
|
|
|||||
Chemical Identifiers
5934-29-2 | |
209.63 | |
CMXXUDSWGMGYLZ-XRIGFGBMSA-N | |
165377 | |
C1=C(NC=N1)CC(C(=O)O)N.O.Cl |
C6H12ClN3O3 | |
MFCD00151027 | |
l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.h2o, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrate | |
(2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid;hydrate;hydrochloride |
Specifications
5934-29-2 | |
C6H12ClN3O3 | |
l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.h2o, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrate | |
C1=C(NC=N1)CC(C(=O)O)N.O.Cl | |
25 g | |
165377 | |
Powder |
White | |
MFCD00151027 | |
CMXXUDSWGMGYLZ-XRIGFGBMSA-N | |
(2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid;hydrate;hydrochloride | |
209.63 | |
209.6 | |
L-Histidine Monohydrochloride Monohydrate |
Safety and Handling
Recommended Storage : Store at Room Temperature (15 to 30°C).