missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals L-Isoleucine, 99%
Supplier: Thermo Scientific Chemicals 166175000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
284.0°C to 286.0°C | |
73-32-5 | |
500ppm max. | |
99% | |
Authentic | |
MFCD00064222 MFCD00004268 | |
04, 454 | |
15, 5225 | |
l-isoleucine, isoleucine, 2s,3s-2-amino-3-methylpentanoic acid, s-isoleucine, s,s-isoleucine, 2s,3s-isoleucine, 2-amino-3-methylvaleric acid, erythro-l-isoleucine, l-+-isoleucine, l-ile | |
AGPKZVBTJJNPAG-WHFBIAKZSA-N | |
(2S,3S)-2-amino-3-methylpentanoic acid | |
+ 41.00 | |
6306 | |
300ppm max. | |
99% | |
+41° (20°C c=4,6N HCl) | |
Crystalline Powder or Crystals |
L-Isoleucine | |
White | |
20ppm max. | |
C6H13NO2 | |
C2H5CH(CH3)CH(NH2)CO2H | |
30ppm max. | |
0.3% max. (105°C, 3 hrs) | |
Plastic Bottle | |
Solubility in water: 41.2g/L (50°C). Other solubilities: soluble in mineral acids, practically insoluble in alcohol, insoluble in ether | |
CC[C@H](C)[C@H](N)C(O)=O | |
0.3% max. | |
131.18 | |
CHEBI:17191 | |
131.17 | |
168.0 °C | |
500 g |
Chemical Identifiers
73-32-5 | |
131.18 | |
AGPKZVBTJJNPAG-WHFBIAKZSA-N | |
6306 | |
(2S,3S)-2-amino-3-methylpentanoic acid |
C6H13NO2 | |
MFCD00064222 MFCD00004268 | |
l-isoleucine, isoleucine, 2s,3s-2-amino-3-methylpentanoic acid, s-isoleucine, s,s-isoleucine, 2s,3s-isoleucine, 2-amino-3-methylvaleric acid, erythro-l-isoleucine, l-+-isoleucine, l-ile | |
CHEBI:17191 | |
CC[C@H](C)[C@H](N)C(O)=O |
RUO – Research Use Only