Learn More
L-Lactide, 98+%, Thermo Scientific Chemicals
$58.68 - $146.00
Chemical Identifiers
CAS | 4511-42-6 |
---|---|
Molecular Formula | C6H8O4 |
Molecular Weight (g/mol) | 144.13 |
MDL Number | MFCD00070594 |
InChI Key | JJTUDXZGHPGLLC-UHFFFAOYNA-N |
Synonym | l-lactide, 3s,6s-3,6-dimethyl-1,4-dioxane-2,5-dione, l---lactide, lactide, l, l-dilactide, 3s-cis-3,6-dimethyl-1,4-dioxane-2,5-dione, ll-lactide, unii-ij13to4no1, 1,4-dioxane-2,5-dione, 3,6-dimethyl-, 3s,6s, ij13to4no1 |
PubChem CID | 107983 |
SMILES | CC1OC(=O)C(C)OC1=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0903106
|
Thermo Scientific Chemicals
L0903106 |
5 g |
Each for $58.68
|
|
|||||
AAL0903114
|
Thermo Scientific Chemicals
L0903114 |
25 g |
Each for $146.00
|
|
|||||
Description
L-Lactide upon polymerization gives poly(lactic acid), which finds many useful properties like biodegradable and nontoxic to the human body and the environment. Its polymer, poly-l-lactide is utilized in the repairing of orbital posterior floor fracture. Further, it is used as biomedical materials for tissue regeneration, matrices for drug delivery systems and alternatives for commercial polymeric materials to reduce the impact on the environment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in chloroform, and methanol. Slightly soluble in benzene.
Notes
Moisture sensitive. Store in a cool place. Incompatible with oxidizing agents.
Chemical Identifiers
4511-42-6 | |
144.13 | |
JJTUDXZGHPGLLC-UHFFFAOYNA-N | |
107983 |
C6H8O4 | |
MFCD00070594 | |
l-lactide, 3s,6s-3,6-dimethyl-1,4-dioxane-2,5-dione, l---lactide, lactide, l, l-dilactide, 3s-cis-3,6-dimethyl-1,4-dioxane-2,5-dione, ll-lactide, unii-ij13to4no1, 1,4-dioxane-2,5-dione, 3,6-dimethyl-, 3s,6s, ij13to4no1 | |
CC1OC(=O)C(C)OC1=O |
Specifications
3.0 | |
4511-42-6 | |
5 g | |
C6H8O4 | |
82124 | |
Soluble in chloroform,and methanol. Slightly soluble in benzene. | |
CC1OC(=O)C(C)OC1=O | |
144.13 | |
144.13 | |
L-Lactide |
95°C to 99°C | |
255°C | |
1.4475 | |
MFCD00070594 | |
l-lactide, 3s,6s-3,6-dimethyl-1,4-dioxane-2,5-dione, l---lactide, lactide, l, l-dilactide, 3s-cis-3,6-dimethyl-1,4-dioxane-2,5-dione, ll-lactide, unii-ij13to4no1, 1,4-dioxane-2,5-dione, 3,6-dimethyl-, 3s,6s, ij13to4no1 | |
JJTUDXZGHPGLLC-UHFFFAOYNA-N | |
3,6-dimethyl-1,4-dioxane-2,5-dione | |
107983 | |
≥98% |
Safety and Handling
GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 224-832-0
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only