missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-leucine, 99.42%, MP Biomedicals™
$803.76 - $803.76
Chemical Identifiers
CAS | 61-90-5 |
---|---|
Molecular Formula | C6H13NO2 |
Molecular Weight (g/mol) | 131.18 |
MDL Number | MFCD00002617 |
InChI Key | ROHFNLRQFUQHCH-MLHKIVSYNA-N |
Synonym | l-leucine, leucine, s-leucine, s-2-amino-4-methylpentanoic acid, h-leu-oh, 2s-2-amino-4-methylpentanoic acid, s-+-leucine, leucin, l-norvaline, 4-methyl, s-2-amino-4-methylvaleric acid |
PubChem CID | 6106 |
ChEBI | CHEBI:15603 |
SMILES | CC(C)C[C@H](N)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN10215801
|
MP Biomedicals Inc
0210215801 |
25 g |
Each for $803.76
|
|
|||||
Chemical Identifiers
61-90-5 | |
131.18 | |
ROHFNLRQFUQHCH-MLHKIVSYNA-N | |
6106 | |
CC(C)C[C@H](N)C(O)=O |
C6H13NO2 | |
MFCD00002617 | |
l-leucine, leucine, s-leucine, s-2-amino-4-methylpentanoic acid, h-leu-oh, 2s-2-amino-4-methylpentanoic acid, s-+-leucine, leucin, l-norvaline, 4-methyl, s-2-amino-4-methylvaleric acid | |
CHEBI:15603 |
Specifications
61-90-5 | |
0.9942 | |
MFCD00002617 | |
ROHFNLRQFUQHCH-MLHKIVSYNA-N | |
(2S)-2-amino-4-methylpentanoic acid | |
6106 | |
131.18 | |
25 g | |
L-Leucine |
White | |
C6H13NO2 | |
l-leucine, leucine, s-leucine, s-2-amino-4-methylpentanoic acid, h-leu-oh, 2s-2-amino-4-methylpentanoic acid, s-+-leucine, leucin, l-norvaline, 4-methyl, s-2-amino-4-methylvaleric acid | |
CC(C)C[C@H](N)C(O)=O | |
131.18 | |
CHEBI:15603 | |
1.293g/mL | |
Small Lustrous Plates or Crystalline Powder |