missing translation for 'onlineSavingsMsg'
Learn More

L-Leucinol, 97%

CAS: 7533-40-6 | C6H15NO | 117.19 g/mol

$151.29 - $522.53

Chemical Identifiers

CAS 7533-40-6
Molecular Formula C6H15NO
Molecular Weight (g/mol) 117.19
MDL Number MFCD00063676
InChI Key VPSSPAXIFBTOHY-LURJTMIESA-N
Synonym l-leucinol, s-+-leucinol, h-leucinol, 2s-2-amino-4-methylpentan-1-ol, l-+-leucinol, l +-leucinol, s-2-amino-4-methylpentan-1-ol, leucinol, s-leucinol, 2s-2-amino-4-methyl-1-pentanol
PubChem CID 111307
IUPAC Name (2S)-2-amino-4-methylpentan-1-ol
SMILES CC(C)C[C@H](N)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2374506
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
B2374506
5 g
Each for $151.29
Only null left
 
AAB2374514
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
B2374514
25 g
Each for $522.53
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7533-40-6
117.19
VPSSPAXIFBTOHY-LURJTMIESA-N
111307
CC(C)C[C@H](N)CO
C6H15NO
MFCD00063676
l-leucinol, s-+-leucinol, h-leucinol, 2s-2-amino-4-methylpentan-1-ol, l-+-leucinol, l +-leucinol, s-2-amino-4-methylpentan-1-ol, leucinol, s-leucinol, 2s-2-amino-4-methyl-1-pentanol
(2S)-2-amino-4-methylpentan-1-ol

Specifications

7533-40-6
0.917
5 g
+1.1° (Neat)
C6H15NO
1719240
VPSSPAXIFBTOHY-LURJTMIESA-N
(2S)-2-amino-4-methylpentan-1-ol
111307
97%
1.451
198°C to 200°C
90°C (194°F)
Air Sensitive
MFCD00063676
l-leucinol, s-+-leucinol, h-leucinol, 2s-2-amino-4-methylpentan-1-ol, l-+-leucinol, l +-leucinol, s-2-amino-4-methylpentan-1-ol, leucinol, s-leucinol, 2s-2-amino-4-methyl-1-pentanol
CC(C)C[C@H](N)CO
117.19
117.19
L-Leucinol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.

P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H227-H302-H314-H335

EINECSNumber : 231-400-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only