missing translation for 'onlineSavingsMsg'
Learn More

L-Lysine acetate, 97%

CAS: 57282-49-2 | C8H18N2O4 | 206.24 g/mol

$145.20 - $1300.82

Chemical Identifiers

CAS 57282-49-2
Molecular Formula C8H18N2O4
Molecular Weight (g/mol) 206.24
MDL Number MFCD00039069
InChI Key RRNJROHIFSLGRA-JEDNCBNOSA-N
Synonym l-lysine acetate, l-lysine acetate salt, lysine acetate, l-lysine monoacetate, unii-ttl6g7liwz, lysine acetate salt, lysine, monoacetate, lysine acetate usp, l-lysine, acetate, ttl6g7liwz
PubChem CID 104152
IUPAC Name acetic acid;(2S)-2,6-diaminohexanoic acid
SMILES CC(O)=O.NCCCC[C@H](N)C(O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC451630250
SDS View Documents
Thermo Scientific Chemicals
451630250
25 g
Each for $145.20
Only null left
 
AC451631000
SDS View Documents
Thermo Scientific Chemicals
451631000
100 g
Each for $388.02
Only null left
 
AC451635000
SDS View Documents
Thermo Scientific Chemicals
451635000
500 g
Each for $1,300.82
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

57282-49-2
206.24
RRNJROHIFSLGRA-JEDNCBNOSA-N
104152
CC(O)=O.NCCCC[C@H](N)C(O)=O
C8H18N2O4
MFCD00039069
l-lysine acetate, l-lysine acetate salt, lysine acetate, l-lysine monoacetate, unii-ttl6g7liwz, lysine acetate salt, lysine, monoacetate, lysine acetate usp, l-lysine, acetate, ttl6g7liwz
acetic acid;(2S)-2,6-diaminohexanoic acid

Specifications

57282-49-2
C8H18N2O4
l-lysine acetate, l-lysine acetate salt, lysine acetate, l-lysine monoacetate, unii-ttl6g7liwz, lysine acetate salt, lysine, monoacetate, lysine acetate usp, l-lysine, acetate, ttl6g7liwz
CC(O)=O.NCCCC[C@H](N)C(O)=O
25 g
104152
97%
97%
MFCD00039069
RRNJROHIFSLGRA-JEDNCBNOSA-N
acetic acid;(2S)-2,6-diaminohexanoic acid
206.24
206.24
L-Lysine acetate
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only