missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Lysine ethyl ester dihydrochloride, 99%, Thermo Scientific Chemicals

$149.10 - $402.44

Chemical Identifiers

CAS 3844-53-9
Molecular Formula C8H20Cl2N2O2
Molecular Weight (g/mol) 247.16
MDL Number MFCD00039068
InChI Key DZIYAIZKJOHVQC-KLXURFKVSA-N
Synonym l-lysine ethyl ester dihydrochloride, h-lys-oet.2hcl, ethyl l-lysinate dihydrochloride, ethyl 2s-2,6-diaminohexanoate dihydrochloride, ethyl l-lysinate hcl, s-ethyl 2,6-diaminohexanoate dihydrochloride, l-lysine, ethyl ester, dihydrochloride, h-lys-oet·2hcl, h-lys-oet inverted exclamation mark currency2hcl, lys-oet2hcl
PubChem CID 107468
IUPAC Name ethyl (2S)-2,6-diaminohexanoate;dihydrochloride
SMILES Cl.Cl.CCOC(=O)[C@@H](N)CCCCN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1682414
View Documents
Thermo Scientific Chemicals
A1682414
25 g
Each for $149.10
 
AAA1682422
View Documents
Thermo Scientific Chemicals
A1682422
100 g
Each for $402.44
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3844-53-9
247.16
DZIYAIZKJOHVQC-KLXURFKVSA-N
107468
Cl.Cl.CCOC(=O)[C@@H](N)CCCCN
C8H20Cl2N2O2
MFCD00039068
l-lysine ethyl ester dihydrochloride, h-lys-oet.2hcl, ethyl l-lysinate dihydrochloride, ethyl 2s-2,6-diaminohexanoate dihydrochloride, ethyl l-lysinate hcl, s-ethyl 2,6-diaminohexanoate dihydrochloride, l-lysine, ethyl ester, dihydrochloride, h-lys-oet·2hcl, h-lys-oet inverted exclamation mark currency2hcl, lys-oet2hcl
ethyl (2S)-2,6-diaminohexanoate;dihydrochloride

Specifications

∼145°C (decomposition)
C8H20Cl2N2O2
3697099
DZIYAIZKJOHVQC-KLXURFKVSA-N
ethyl (2S)-2,6-diaminohexanoate;dihydrochloride
247.16
247.17
99%
3844-53-9
MFCD00039068
l-lysine ethyl ester dihydrochloride, h-lys-oet.2hcl, ethyl l-lysinate dihydrochloride, ethyl 2s-2,6-diaminohexanoate dihydrochloride, ethyl l-lysinate hcl, s-ethyl 2,6-diaminohexanoate dihydrochloride, l-lysine, ethyl ester, dihydrochloride, h-lys-oet·2hcl, h-lys-oet inverted exclamation mark currency2hcl, lys-oet2hcl
Cl.Cl.CCOC(=O)[C@@H](N)CCCCN
25 g
107468
Hygroscopic
L-Lysine ethyl ester dihydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 223-340-3

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only