missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Lysine ethyl ester diisocyanate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6438414

 View more versions of this product

Catalog No. AAH6438414


Explore available promotions

Description

Description

It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Sparingly soluble in water (0.19 g/L).

Notes
Moisture Sensitive. Store away form water/moisture. Store away from oxidizing agent.
Specifications

Specifications

L-Lysine ethyl ester diisocyanate
Yellow to Brown
1.4572
s-ethyl 2,6-diisocyanatohexanoate, l-lysine ethyl ester diisocyanate, ethyllysine diisocyanate, ethyllysine diisocyanate;, ksc005q3n, lysine ethyl ester diisocyanate;, l-lysine diisocyanate ethyl ester, l-lysine ethyl ester diisocyanate;, ethyl 2s-2,6-diisocyanatohexanoate, s-2,6-diisocyanatohexanoic acid ethyl ester
IGWTZHMYJZZIGC-IMWMWJONNA-N
ethyl (2S)-2,6-diisocyanatohexanoate
25 g
226.23
Moisture sensitive
45172-15-4
C10H14N2O4
MFCD08276480
Sparingly soluble in water (0.19g/L).
CCOC(=O)[C@H](CCCCN=C=O)N=C=O
226.23
71440643
97%
Viscous Liquid

Chemical Identifiers

45172-15-4
226.23
IGWTZHMYJZZIGC-IMWMWJONNA-N
71440643
CCOC(=O)[C@H](CCCCN=C=O)N=C=O
C10H14N2O4
MFCD08276480
s-ethyl 2,6-diisocyanatohexanoate, l-lysine ethyl ester diisocyanate, ethyllysine diisocyanate, ethyllysine diisocyanate;, ksc005q3n, lysine ethyl ester diisocyanate;, l-lysine diisocyanate ethyl ester, l-lysine ethyl ester diisocyanate;, ethyl 2s-2,6-diisocyanatohexanoate, s-2,6-diisocyanatohexanoic acid ethyl ester
ethyl (2S)-2,6-diisocyanatohexanoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H334-H315-H319-H317-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P311-P333+P313-P362-P501c

H315-H317-H319-H331-H334-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only