Learn More
L-Lysine monohydrate, 97%, Thermo Scientific Chemicals
$100.66 - $1326.00
Chemical Identifiers
CAS | 39665-12-8 |
---|---|
Molecular Formula | C6H16N2O3 |
Molecular Weight (g/mol) | 164.21 |
MDL Number | MFCD00151035 |
InChI Key | HZRUTVAFDWTKGD-XZNNNFJINA-N |
Synonym | l-lysine monohydrate, l-lysine hydrate, lysine monohydrate, l +-lysine monohydrate, l-lysine, hydrate, h-lys-oh h2o, lysine, hydrate, lysine hydrate, l-lysine, monohydrate, l-lysine, hydrate 1:1 |
PubChem CID | 16211825 |
SMILES | O.NCCCC[C@H](N)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1304014
|
Thermo Scientific Chemicals
A1304014 |
25 g |
Each for $100.66
|
|
|||||
AAA1304022
|
Thermo Scientific Chemicals
A1304022 |
100 g |
Each for $284.68
|
|
|||||
AAA1304036
|
Thermo Scientific Chemicals
A1304036 |
500 g |
Each for $1,326.00
|
|
|||||
Description
L-Lysine is an essential proteinogenic alfa amino acid used in various applications such as non-animal sourced supplement in cell culture media. It acts as a substrate of enzymes like L-lysine oxidase as well as a component of poly-lysine polymers. It is also involved in the studies of glycation mechanism.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsL-Lysine is an essential proteinogenic alfa amino acid used in various applications such as non-animal sourced supplement in cell culture media. It acts as a substrate of enzymes like L-lysine oxidase as well as a component of poly-lysine polymers. It is also involved in the studies of glycation mechanism.
Solubility
Soluble in water and hydrochloric acid.
Notes
Hygroscopic. Air sensitive. Incompatible with strong oxidizing agents.
Chemical Identifiers
39665-12-8 | |
164.21 | |
HZRUTVAFDWTKGD-XZNNNFJINA-N | |
16211825 |
C6H16N2O3 | |
MFCD00151035 | |
l-lysine monohydrate, l-lysine hydrate, lysine monohydrate, l +-lysine monohydrate, l-lysine, hydrate, h-lys-oh h2o, lysine, hydrate, lysine hydrate, l-lysine, monohydrate, l-lysine, hydrate 1:1 | |
O.NCCCC[C@H](N)C(O)=O |
Specifications
210°C (decomposition) | |
C6H16N2O3 | |
1722531 | |
l-lysine monohydrate, l-lysine hydrate, lysine monohydrate, l +-lysine monohydrate, l-lysine, hydrate, h-lys-oh h2o, lysine, hydrate, lysine hydrate, l-lysine, monohydrate, l-lysine, hydrate 1:1 | |
HZRUTVAFDWTKGD-XZNNNFJINA-N | |
2,6-diaminohexanoic acid hydrate | |
16211825 | |
97% | |
+22.5° (c=2 in 6M HCl) | |
L-Lysine monohydrate |
39665-12-8 | |
MFCD00151035 | |
14,5636 | |
Soluble in water and hydrochloric acid. | |
O.NCCCC[C@H](N)C(O)=O | |
164.21 | |
164.21 (146.19 Anhydrous) | |
25 g | |
Air sensitive; Hygroscopic |
Safety and Handling
EINECSNumber : 200-294-2
RTECSNumber : OL5540000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only