missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals L-Lysine monohydrochloride, Cell Culture Reagent

Supplier:  Thermo Scientific Chemicals J62099A1

 View more versions of this product

Catalog No. AAJ62099A1


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

L-Lysine monohydrochloride
White
MFCD00064564 MFCD00081870
14,5636
BVHLGVCQOALMSV-JEDNCBNOSA-N
(2S)-2,6-diaminohexanoic acid;hydrochloride
182.65
CHEBI:53633
Cell Culture Reagent
657-27-2
C6H15ClN2O2
3563889
l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisyl
Cl.NCCCC[C@H](N)C(O)=O
1 kg
69568
182.65
Powder

Chemical Identifiers

657-27-2
182.65
BVHLGVCQOALMSV-JEDNCBNOSA-N
69568
(2S)-2,6-diaminohexanoic acid;hydrochloride
C6H15ClN2O2
MFCD00064564 MFCD00081870
l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisyl
CHEBI:53633
Cl.NCCCC[C@H](N)C(O)=O
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 211-519-9

RTECSNumber : OL5650000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only