missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-(-)-Malic acid, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals J6322136

 View more versions of this product

Catalog No. AAJ6322136


Explore available promotions

Description

Description

L-(-)-Malic acid is used as a food additive, Selective α-amino protecting reagent for amino acid derivatives. Versatile synthon for the preparation of chiral compounds including κ-opioid receptor agonists, 1α,25-dihydroxyvitamin D3 analogue, and phoslactomycin B.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-(-)-Malic acid is used as a food additive, Selective α-amino protecting reagent for amino acid derivatives. Versatile synthon for the preparation of chiral compounds including κ-opioid receptor agonists, 1α,25-dihydroxyvitamin D3 analogue, and phoslactomycin B.

Solubility
Soluble in water(363g/L).

Notes
Keep container tightly closed. Store away from oxidizing agents. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

L-(-)-Malic acid
100°C to 106°C
1.6
C4H6O5
500 g
14,5707
Soluble in water(363g/L).
−26° (c=5.5 in pyridine)
(2S)-2-hydroxybutanedioic acid
222656
134.09
Crystalline Powder
97-67-6
White
220°C (428°F)
MFCD00064213
1723541
l-malic acid, l---malic acid, s-2-hydroxysuccinic acid, 2s-2-hydroxybutanedioic acid, l--malic acid, apple acid, --malic acid, l-apple acid, s-malic acid, s-2-hydroxybutanedioic acid
BJEPYKJPYRNKOW-REOHCLBHSA-N
C(C(C(=O)O)O)C(=O)O
134.087
CHEBI:30797
99%

Chemical Identifiers

97-67-6
134.087
BJEPYKJPYRNKOW-REOHCLBHSA-N
222656
(2S)-2-hydroxybutanedioic acid
C4H6O5
MFCD00064213
l-malic acid, l---malic acid, s-2-hydroxysuccinic acid, 2s-2-hydroxybutanedioic acid, l--malic acid, apple acid, --malic acid, l-apple acid, s-malic acid, s-2-hydroxybutanedioic acid
CHEBI:30797
C(C(C(=O)O)O)C(=O)O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c

H315-H318-H335

EINECSNumber : 202-601-5

RTECSNumber : ON7175000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only