missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Menthol, Crystal, USP, 98-102%, Spectrum™ Chemical
ME110, 2216-51-5, C10H20O
Supplier: Spectrum Chemical Mfg Cor ME11025KGBL
Description
Spectrum™ Chemical L-Menthol, Crystal, USP is used as a local anesthetic, a counterirritant and can help relieve minor throat irritations. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
2216-51-5 | |
41-44°C | |
C10H20O | |
-45° to -51° | |
CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O | |
156.27 | |
USP |
100% | |
Fiber Drum | |
25 kg | |
NOOLISFMXDJSKH-KXUCPTDWSA-N | |
(1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol | |
98 to 102% |
Chemical Identifiers
2216-51-5 | |
156.27 | |
(1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol |
C10H20O | |
NOOLISFMXDJSKH-KXUCPTDWSA-N | |
CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O |