Learn More
l-Menthone, 85%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 203610250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
l-Menthone | |
14073-97-3 | |
0.8900g/mL | |
72°C | |
80% min. (GC) | |
C10H18O | |
MFCD00001634,MFCD00136033 | |
07, 38 | |
0.89 | |
−6° (20°C c=4,ethanol) | |
--menthone, l-menthone, menthone, trans-menthone, p-menthone, neomenthone, menthone racemic, trans-p-menthan-3-one, 2s,5r-2-isopropyl-5-methylcyclohexanone, l-menthan-3-one | |
NFLGAXVYCFJBMK-BDAKNGLRSA-N | |
(2S,5R)-5-methyl-2-propan-2-ylcyclohexan-1-one | |
26447 | |
154.25 | |
Liquid |
85% | |
Colorless to Yellow | |
207°C to 210°C | |
Authentic | |
Glass bottle | |
1.4490 to 1.453 | |
25 mL | |
14,201; 16,204 | |
15, 5906 | |
-6 | |
Solubility in water: slightly soluble. Other solubilities: soluble in acetone,miscible with ethanol and,ethyl ether | |
CC(C)[C@@H]1CC[C@@H](C)CC1=O | |
154.25 | |
CHEBI:15410 | |
85% |
Chemical Identifiers
14073-97-3 | |
154.25 | |
NFLGAXVYCFJBMK-BDAKNGLRSA-N | |
26447 | |
(2S,5R)-5-methyl-2-propan-2-ylcyclohexan-1-one |
C10H18O | |
MFCD00001634,MFCD00136033 | |
--menthone, l-menthone, menthone, trans-menthone, p-menthone, neomenthone, menthone racemic, trans-p-menthan-3-one, 2s,5r-2-isopropyl-5-methylcyclohexanone, l-menthan-3-one | |
CHEBI:15410 | |
CC(C)[C@@H]1CC[C@@H](C)CC1=O |
Safety and Handling
GHS H Statement:
Harmful to aquatic life with long lasting effects.
GHS P Statement:
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 237-926-1
TSCA : TSCA
RUO – Research Use Only