Learn More
L-Menthone, 97%
CAS: 14073-97-3 | C10H18O | 154.25 g/mol
$43.78 - $407.78
Chemical Identifiers
| CAS | 14073-97-3 |
|---|---|
| Molecular Formula | C10H18O |
| Molecular Weight (g/mol) | 154.25 |
| MDL Number | MFCD00001634,MFCD00136033 |
| InChI Key | NFLGAXVYCFJBMK-BDAKNGLRSA-N |
| Synonym | --menthone, l-menthone, menthone, trans-menthone, p-menthone, neomenthone, menthone racemic, trans-p-menthan-3-one, 2s,5r-2-isopropyl-5-methylcyclohexanone, l-menthan-3-one |
| PubChem CID | 26447 |
| ChEBI | CHEBI:15410 |
| IUPAC Name | (2S,5R)-5-methyl-2-propan-2-ylcyclohexan-1-one |
| SMILES | CC(C)[C@@H]1CC[C@@H](C)CC1=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1367909
|
Thermo Scientific Chemicals
A1367909 |
10 g |
Each for $43.78
|
|
|||||
|
AAA1367918
|
Thermo Scientific Chemicals
A1367918 |
50 g |
Each for $106.97
|
|
|||||
|
AAA1367922
|
Thermo Scientific Chemicals
A1367922 |
100 g |
Each for $199.48
|
|
|||||
|
AAA1367930
|
Thermo Scientific Chemicals
A1367930 |
250 g |
Each for $407.78
|
|
|||||
Description
L-Menthone is used in perfumery and cosmetics for its characteristic aromatic and minty odor. And it is also used in synthesis of agrochemicals, and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsL-Menthone is used in perfumery and cosmetics for its characteristic aromatic and minty odor. And it is also used in synthesis of agrochemicals, and dyestuff.
Solubility
Soluble in ether, alcohol, benzene, acetone.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
| 14073-97-3 | |
| 154.25 | |
| NFLGAXVYCFJBMK-BDAKNGLRSA-N | |
| 26447 | |
| (2S,5R)-5-methyl-2-propan-2-ylcyclohexan-1-one |
| C10H18O | |
| MFCD00001634,MFCD00136033 | |
| --menthone, l-menthone, menthone, trans-menthone, p-menthone, neomenthone, menthone racemic, trans-p-menthan-3-one, 2s,5r-2-isopropyl-5-methylcyclohexanone, l-menthan-3-one | |
| CHEBI:15410 | |
| CC(C)[C@@H]1CC[C@@H](C)CC1=O |
Specifications
| 14073-97-3 | |
| 208°C to 210°C | |
| 97% | |
| 1.45 | |
| 10 g | |
| 14,5838 | |
| Soluble in ether,alcohol,benzene,acetone. | |
| −25° (Neat) | |
| (2S,5R)-5-methyl-2-propan-2-ylcyclohexan-1-one | |
| 26447 | |
| 154.25 | |
| Liquid |
| 0.893 | |
| 72°C (162°F) | |
| C10H18O | |
| MFCD00001634,MFCD00136033 | |
| 3648743 | |
| --menthone, l-menthone, menthone, trans-menthone, p-menthone, neomenthone, menthone racemic, trans-p-menthan-3-one, 2s,5r-2-isopropyl-5-methylcyclohexanone, l-menthan-3-one | |
| NFLGAXVYCFJBMK-BDAKNGLRSA-N | |
| CC(C)[C@@H]1CC[C@@H](C)CC1=O | |
| 154.25 | |
| CHEBI:15410 | |
| 97% | |
| L-Menthone |
Safety and Handling
GHS H Statement
H319-H227
Causes serious eye irritation.
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 237-926-1
RTECSNumber : OS9542200
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only