missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals L-Methionine sulfone, 98+%
$89.32 - $284.89
Chemical Identifiers
CAS | 7314-32-1 |
---|---|
Molecular Formula | C5H11NO4S |
Molecular Weight (g/mol) | 181.21 |
MDL Number | MFCD00066020 |
InChI Key | UCUNFLYVYCGDHP-BYPYZUCNSA-N |
Synonym | l-methionine sulfone, s-2-amino-4-methylsulfonyl butanoic acid, s-dioxymethionine, l-2-amino-4-methylsulfonyl butanoic acid, 2s-2-amino-4-methylsulfonylbutanoic acid, 2s-2-amino-4-methanesulfonylbutanoic acid, methionine s,s-dioxide, s-amino-4-methylsulphonyl butyric acid, h-met o 2-oh, l-methioninesulfone,99+% |
PubChem CID | 445282 |
ChEBI | CHEBI:21363 |
IUPAC Name | (2S)-2-amino-4-methylsulfonylbutanoic acid |
SMILES | CS(=O)(=O)CC[C@H](N)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1702703
|
Thermo Scientific Chemicals
A1702703 |
1 g |
Each for $89.32
|
|
|||||
AAA1702706
|
Thermo Scientific Chemicals
A1702706 |
5 g |
Each for $284.89
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7314-32-1 | |
181.21 | |
UCUNFLYVYCGDHP-BYPYZUCNSA-N | |
445282 | |
(2S)-2-amino-4-methylsulfonylbutanoic acid |
C5H11NO4S | |
MFCD00066020 | |
l-methionine sulfone, s-2-amino-4-methylsulfonyl butanoic acid, s-dioxymethionine, l-2-amino-4-methylsulfonyl butanoic acid, 2s-2-amino-4-methylsulfonylbutanoic acid, 2s-2-amino-4-methanesulfonylbutanoic acid, methionine s,s-dioxide, s-amino-4-methylsulphonyl butyric acid, h-met o 2-oh, l-methioninesulfone,99+% | |
CHEBI:21363 | |
CS(=O)(=O)CC[C@H](N)C(O)=O |
Specifications
254°C (decomposition) | |
C5H11NO4S | |
1725510 | |
UCUNFLYVYCGDHP-BYPYZUCNSA-N | |
(2S)-2-amino-4-methylsulfonylbutanoic acid | |
181.21 | |
CHEBI:21363 | |
181.21 | |
L-Methionine sulfone |
7314-32-1 | |
MFCD00066020 | |
l-methionine sulfone, s-2-amino-4-methylsulfonyl butanoic acid, s-dioxymethionine, l-2-amino-4-methylsulfonyl butanoic acid, 2s-2-amino-4-methylsulfonylbutanoic acid, 2s-2-amino-4-methanesulfonylbutanoic acid, methionine s,s-dioxide, s-amino-4-methylsulphonyl butyric acid, h-met o 2-oh, l-methioninesulfone,99+% | |
CS(=O)(=O)CC[C@H](N)C(O)=O | |
1 g | |
445282 | |
+11.6° (c=0.69 in Water) | |
≥99% |
Safety and Handling
EINECSNumber : 230-774-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only