missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Norvaline, 99%

CAS: 6600-40-4 | C5H11NO2 | 117.15 g/mol

$64.44 - $174.05

Chemical Identifiers

CAS 6600-40-4
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00064421
InChI Key SNDPXSYFESPGGJ-BYPYZUCNSA-N
Synonym l-norvaline, norvaline, s-2-aminopentanoic acid, h-nva-oh, 2s-2-aminopentanoic acid, l-2-aminovaleric acid, l-2-aminopentanoic acid, norvaline, l, s-2-aminovaleric acid, l-+-norvaline
PubChem CID 65098
ChEBI CHEBI:18314
IUPAC Name (2S)-2-aminopentanoic acid
SMILES CCC[C@H](N)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0865803
View Documents
Thermo Scientific Chemicals
L0865803
1 g
Each for $64.44
Only null left
 
AAL0865806
View Documents
Thermo Scientific Chemicals
L0865806
5 g
Each for $174.05
Only null left
 
Description

Description

L-Norvaline enhances NO production from activated macrophages. It is known to promote tissue regeneration and muscle growth and become a precursor in the penicillin biosynthetic pathway. It also can be used for nutrition agent and drug synthesis. For example: it can react with phthalic acid anhydride to get N,N-phthaloyl-L-norvaline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Norvaline enhances NO production from activated macrophages. It is known to promote tissue regeneration and muscle growth and become a precursor in the penicillin biosynthetic pathway. It also can be used for nutrition agent and drug synthesis. For example: it can react with phthalic acid anhydride to get N,N-phthaloyl-L-norvaline.

Solubility
Soluble in water (almost transparency), 10.5 g/100 mL (18°C).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

6600-40-4
117.15
SNDPXSYFESPGGJ-BYPYZUCNSA-N
65098
(2S)-2-aminopentanoic acid
C5H11NO2
MFCD00064421
l-norvaline, norvaline, s-2-aminopentanoic acid, h-nva-oh, 2s-2-aminopentanoic acid, l-2-aminovaleric acid, l-2-aminopentanoic acid, norvaline, l, s-2-aminovaleric acid, l-+-norvaline
CHEBI:18314
CCC[C@H](N)C(O)=O

Specifications

>300°C
C5H11NO2
1721162
l-norvaline, norvaline, s-2-aminopentanoic acid, h-nva-oh, 2s-2-aminopentanoic acid, l-2-aminovaleric acid, l-2-aminopentanoic acid, norvaline, l, s-2-aminovaleric acid, l-+-norvaline
SNDPXSYFESPGGJ-BYPYZUCNSA-N
(2S)-2-aminopentanoic acid
117.15
CHEBI:18314
117.15
L-Norvaline
6600-40-4
MFCD00064421
14,6716
Soluble in water (almost transparency),10.5g/100 mL (18°C).
CCC[C@H](N)C(O)=O
1 g
65098
+25° (c=10 in 20% HCl)
99%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 229-543-3

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only