missing translation for 'onlineSavingsMsg'
Learn More

L-Norvaline Ethyl Ester Hydrochloride 98.0+%, TCI America™

$67.11 - $206.90

Chemical Identifiers

CAS 40918-51-2
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD08458628
InChI Key CHAJBHNQIZAJJM-UHFFFAOYNA-N
Synonym (S)-2-Aminopentanoic Acid Ethyl Ester Hydrochloride, (S)-2-Aminovaleric Acid Ethyl Ester Hydrochloride, Ethyl (S)-2-Aminopentanoate Hydrochloride, Ethyl (S)-2-Aminovalerate Hydrochloride, H-Nva-OEt.HCl
PubChem CID 12631224
IUPAC Name ethyl 2-aminopentanoate hydrochloride
SMILES Cl.CCCC(N)C(=O)OCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N08985G
View Documents
TCI America
N08985G
5 g
Each for $67.11
Only null left
 
N089825G
View Documents
TCI America
N089825G
25 g
Each for $206.90
Only null left
 
Description

Description

Specifications

Chemical Identifiers

40918-51-2
181.66
CHAJBHNQIZAJJM-UHFFFAOYNA-N
12631224
Cl.CCCC(N)C(=O)OCC
C7H16ClNO2
MFCD08458628
(S)-2-Aminopentanoic Acid Ethyl Ester Hydrochloride, (S)-2-Aminovaleric Acid Ethyl Ester Hydrochloride, Ethyl (S)-2-Aminopentanoate Hydrochloride, Ethyl (S)-2-Aminovalerate Hydrochloride, H-Nva-OEt.HCl
ethyl 2-aminopentanoate hydrochloride

Specifications

40918-51-2
White
MFCD08458628
(S)-2-Aminopentanoic Acid Ethyl Ester Hydrochloride, (S)-2-Aminovaleric Acid Ethyl Ester Hydrochloride, Ethyl (S)-2-Aminopentanoate Hydrochloride, Ethyl (S)-2-Aminovalerate Hydrochloride, H-Nva-OEt.HCl
Cl.CCCC(N)C(=O)OCC
181.66
181.66
Crystalline Powder
109°C
C7H16ClNO2
5 g
CHAJBHNQIZAJJM-UHFFFAOYNA-N
ethyl 2-aminopentanoate hydrochloride
12631224
≥98.0% (N,T)
L-Norvaline Ethyl Ester Hydrochloride
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'