missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L(+)-Ornithine hydrochloride, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 129561000

 View more versions of this product

Catalog No. AC129561000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

L(+)-Ornithine hydrochloride
245.0°C
Authentic
Plastic bottle
H2N(CH2)3CH(NH2)CO2H·HCl
100 g
14, 6874
+ 24.00
(5% in 1 N HCl) Clear colorless
C(CC(C(=O)O)N)CN.Cl
168.62
168.62
Powder
3184-13-2
White
98.5% min. (Argentometry)
C5H12N2O2·HCl
MFCD00064562
04, 420
+ 24.00 (20.00°C c=4,6N HCl)
l-ornithine hydrochloride, l-ornithine monohydrochloride, h-orn-oh.hcl, l-ornithine hcl, l +-ornithine hydrochloride, l-ornithine, monohydrochloride, s-2,5-diaminopentanoic acid hydrochloride, ornithine hydrochloride van, unii-hbk84k66xh, ornithine l monochlorohydrate
GGTYBZJRPHEQDG-WCCKRBBISA-N
(2S)-2,5-diaminopentanoic acid;hydrochloride
76654
99%

Chemical Identifiers

3184-13-2
168.62
GGTYBZJRPHEQDG-WCCKRBBISA-N
76654
C(CC(C(=O)O)N)CN.Cl
C5H12N2O2·HCl
MFCD00064562
l-ornithine hydrochloride, l-ornithine monohydrochloride, h-orn-oh.hcl, l-ornithine hcl, l +-ornithine hydrochloride, l-ornithine, monohydrochloride, s-2,5-diaminopentanoic acid hydrochloride, ornithine hydrochloride van, unii-hbk84k66xh, ornithine l monochlorohydrate
(2S)-2,5-diaminopentanoic acid;hydrochloride
Safety and Handling

Safety and Handling

EINECSNumber : 221-678-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only