Learn More
L-Phenylalanine ethyl ester hydrochloride, 98%
CAS: 3182-93-2 | C11H16ClNO2 | 229.70 g/mol
$60.90 - $626.68
Chemical Identifiers
| CAS | 3182-93-2 |
|---|---|
| Molecular Formula | C11H16ClNO2 |
| Molecular Weight (g/mol) | 229.70 |
| MDL Number | MFCD00012507 |
| InChI Key | FPFQPLFYTKMCHN-PPHPATTJSA-N |
| Synonym | l-phenylalanine ethyl ester hydrochloride, h-phe-oet.hcl, ethyl l-phenylalaninate hydrochloride, s-ethyl 2-amino-3-phenylpropanoate hydrochloride, ethyl 2s-2-amino-3-phenylpropanoate hydrochloride, l-phenylalanine, ethyl ester, hydrochloride, l-phenylalanine ethylester, ethyl l-phenylalaninate hcl, phenylalanine ethyl ester hydrochloride, phe-oet.hcl |
| PubChem CID | 165085 |
| IUPAC Name | ethyl (2S)-2-amino-3-phenylpropanoate;hydrochloride |
| SMILES | [H+].[Cl-].CCOC(=O)[C@@H](N)CC1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1361806
|
Thermo Scientific Chemicals
A1361806 |
5 g |
Each for $60.90
|
|
|||||
|
AAA1361814
|
Thermo Scientific Chemicals
A1361814 |
25 g |
Each for $183.59
|
|
|||||
|
AAA1361822
|
Thermo Scientific Chemicals
A1361822 |
100 g |
Each for $626.68
|
|
|||||
Description
L-Phenylalanine ethyl ester hydrochloride is used as an enzyme substrate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsL-Phenylalanine ethyl ester hydrochloride is used as an enzyme substrate.
Solubility
Soluble in methanol and water.
Notes
Keep container tightly sealed. Incompatible with oxidizing agents. Store under dry inert gas. It is hygroscopic in nature. Protect from humidity and water.
Chemical Identifiers
| 3182-93-2 | |
| 229.70 | |
| FPFQPLFYTKMCHN-PPHPATTJSA-N | |
| 165085 | |
| [H+].[Cl-].CCOC(=O)[C@@H](N)CC1=CC=CC=C1 |
| C11H16ClNO2 | |
| MFCD00012507 | |
| l-phenylalanine ethyl ester hydrochloride, h-phe-oet.hcl, ethyl l-phenylalaninate hydrochloride, s-ethyl 2-amino-3-phenylpropanoate hydrochloride, ethyl 2s-2-amino-3-phenylpropanoate hydrochloride, l-phenylalanine, ethyl ester, hydrochloride, l-phenylalanine ethylester, ethyl l-phenylalaninate hcl, phenylalanine ethyl ester hydrochloride, phe-oet.hcl | |
| ethyl (2S)-2-amino-3-phenylpropanoate;hydrochloride |
Specifications
| 153°C to 157°C | |
| C11H16ClNO2 | |
| 3657823 | |
| Soluble in methanol and water. | |
| [H+].[Cl-].CCOC(=O)[C@@H](N)CC1=CC=CC=C1 | |
| 5 g | |
| 165085 | |
| 229.71 | |
| 98% |
| 3182-93-2 | |
| MFCD00012507 | |
| l-phenylalanine ethyl ester hydrochloride, h-phe-oet.hcl, ethyl l-phenylalaninate hydrochloride, s-ethyl 2-amino-3-phenylpropanoate hydrochloride, ethyl 2s-2-amino-3-phenylpropanoate hydrochloride, l-phenylalanine, ethyl ester, hydrochloride, l-phenylalanine ethylester, ethyl l-phenylalaninate hcl, phenylalanine ethyl ester hydrochloride, phe-oet.hcl | |
| FPFQPLFYTKMCHN-PPHPATTJSA-N | |
| ethyl (2S)-2-amino-3-phenylpropanoate;hydrochloride | |
| 229.70 | |
| +36° (c=5 in Ethanol) | |
| Hygroscopic | |
| L-Phenylalanine ethyl ester hydrochloride |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 221-673-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only