missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L(-)-Phenylephrine hydrochloride, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 207240100

 View more versions of this product

Catalog No. AC207240100


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

L(-)-Phenylephrine hydrochloride
140°C to 145°C
1% max. (1 g, 105°C)
99%
C9H14ClNO2
MFCD00012605,MFCD00044749
15, 7398
-47
OCYSGIYOVXAGKQ-UHFFFAOYNA-N
3-[(1R)-1-hydroxy-2-(methylamino)ethyl]phenol;hydrochloride
5284443
203.67
Crystalline Powder
61-76-7
White
Authentic
Glass bottle
HOC6H4CH(CH2NHCH3)OH·HCl
10 g
−47° (20°C c=2,H2O)
phenylephrine hydrochloride, phenylephrine hcl, l-phenylephrine hydrochloride, metaoxedrine chloride, neosympatol, synethenate, almefrin, consdrin, emagrin, fenilfar
[H+].[Cl-].CNCC(O)C1=CC=CC(O)=C1
203.67
CHEBI:8094
99%

Chemical Identifiers

61-76-7
203.67
OCYSGIYOVXAGKQ-UHFFFAOYNA-N
5284443
3-[(1R)-1-hydroxy-2-(methylamino)ethyl]phenol;hydrochloride
C9H14ClNO2
MFCD00012605,MFCD00044749
phenylephrine hydrochloride, phenylephrine hcl, l-phenylephrine hydrochloride, metaoxedrine chloride, neosympatol, synethenate, almefrin, consdrin, emagrin, fenilfar
CHEBI:8094
[H+].[Cl-].CNCC(O)C1=CC=CC(O)=C1
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 200-517-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only