missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Prolinamide, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 302990050

 View more versions of this product

Catalog No. AC302990050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

L-Prolinamide
97.5% min. (TLC)
98%
Glass bottle
− 106.00
VLJNHYLEOZPXFW-BYPYZUCNSA-O
(2S)-pyrrolidine-2-carboxamide
5g
111306
114.15
7531-52-4
Authentic
C5H11N2O
MFCD00005253
l-prolinamide, prolinamide, h-pro-nh2, s-pyrrolidine-2-carboxamide, s-prolinamide, 2s-pyrrolidine-2-carboxamide, l-prolineamide, l-proline amide, l---prolinamide, s-2-pyrrolidinecarboxamide
NC(=O)[C@@H]1CCC[NH2+]1
−106° (20°C c=2,EtOH)
115.16
CHEBI:21374
98%

Chemical Identifiers

7531-52-4
115.16
VLJNHYLEOZPXFW-BYPYZUCNSA-O
111306
(2S)-pyrrolidine-2-carboxamide
C5H11N2O
MFCD00005253
l-prolinamide, prolinamide, h-pro-nh2, s-pyrrolidine-2-carboxamide, s-prolinamide, 2s-pyrrolidine-2-carboxamide, l-prolineamide, l-proline amide, l---prolinamide, s-2-pyrrolidinecarboxamide
CHEBI:21374
NC(=O)[C@@H]1CCC[NH2+]1
Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only