Learn More
L-Proline, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1019936
Description
L-Proline is used as asymmetric catalysts in organic synthesis and asymmetric aldol cyclization. It is involved in the Michael addition of dimethyl malonate to alfa-beta-unsaturated aldehydes. It is a precursor of hydroxyproline in collagen. It is an active component of collagen and involved in the proper functioning of joints and tendons. It finds uses in pharmaceutical, biotechnological applications due to its osmoprotectant property. Further, it is used with ninhydrin in the chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsL-Proline is used as asymmetric catalysts in organic synthesis and asymmetric aldol cyclization. It is involved in the Michael addition of dimethyl malonate to alfa-beta-unsaturated aldehydes. It is a precursor of hydroxyproline in
Specifications
∼225°C (decomposition) | |
147-85-3 | |
MFCD00064318 | |
14,7780 | |
Soluble in water and ethanol. Insoluble in ether,butanol and isopropanol. | |
OC(=O)C1CCCN1 | |
115.13 | |
CHEBI:17203 | |
99% | |
500 g | |
Hygroscopic |
L-Proline | |
C5H9NO2 | |
80810 | |
l-proline, proline, l---proline, s-pyrrolidine-2-carboxylic acid, 2-pyrrolidinecarboxylic acid, --proline, --s-proline, 2s-pyrrolidine-2-carboxylic acid, prolinum, h-pro-oh | |
ONIBWKKTOPOVIA-UHFFFAOYNA-N | |
(2S)-pyrrolidine-2-carboxylic acid | |
145742 | |
115.13 | |
1.36 | |
−84° (c=1 in water) |
Chemical Identifiers
147-85-3 | |
115.13 | |
ONIBWKKTOPOVIA-UHFFFAOYNA-N | |
145742 | |
(2S)-pyrrolidine-2-carboxylic acid |
C5H9NO2 | |
MFCD00064318 | |
l-proline, proline, l---proline, s-pyrrolidine-2-carboxylic acid, 2-pyrrolidinecarboxylic acid, --proline, --s-proline, 2s-pyrrolidine-2-carboxylic acid, prolinum, h-pro-oh | |
CHEBI:17203 | |
OC(=O)C1CCCN1 |
Safety and Handling
EINECSNumber : 205-702-2
RTECSNumber : TW3584000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only