missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Proline, >99%, MP Biomedicals™
$137.39 - $976.75
Chemical Identifiers
CAS | 147-85-3 |
---|---|
Molecular Formula | C5H9NO2 |
Molecular Weight (g/mol) | 115.13 |
MDL Number | MFCD00064318 |
InChI Key | ONIBWKKTOPOVIA-UHFFFAOYNA-N |
Synonym | l-proline, proline, l---proline, s-pyrrolidine-2-carboxylic acid, 2-pyrrolidinecarboxylic acid, --proline, --s-proline, 2s-pyrrolidine-2-carboxylic acid, prolinum, h-pro-oh |
PubChem CID | 145742 |
ChEBI | CHEBI:17203 |
IUPAC Name | pyrrolidine-2-carboxylic acid |
SMILES | OC(=O)C1CCCN1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN10273080
|
MP Biomedicals Inc
0210273080 |
100 g |
Each for $137.39
|
|
|||||
ICN10273091
|
MP Biomedicals Inc
0210273091 |
1 kg |
Each for $976.75
|
|
|||||
Chemical Identifiers
147-85-3 | |
115.13 | |
ONIBWKKTOPOVIA-UHFFFAOYNA-N | |
145742 | |
pyrrolidine-2-carboxylic acid |
C5H9NO2 | |
MFCD00064318 | |
l-proline, proline, l---proline, s-pyrrolidine-2-carboxylic acid, 2-pyrrolidinecarboxylic acid, --proline, --s-proline, 2s-pyrrolidine-2-carboxylic acid, prolinum, h-pro-oh | |
CHEBI:17203 | |
OC(=O)C1CCCN1 |
Specifications
147-85-3 | |
>99% | |
MFCD00064318 | |
ONIBWKKTOPOVIA-UHFFFAOYNA-N | |
pyrrolidine-2-carboxylic acid | |
115.13 | |
CHEBI:17203 | |
>99% | |
L-Proline, Cell Biology and Analysis |
White | |
C5H9NO2 | |
l-proline, proline, l---proline, s-pyrrolidine-2-carboxylic acid, 2-pyrrolidinecarboxylic acid, --proline, --s-proline, 2s-pyrrolidine-2-carboxylic acid, prolinum, h-pro-oh | |
OC(=O)C1CCCN1 | |
100 g | |
145742 | |
115.13 | |
Powder |
Safety and Handling
EINECSNumber : 205-702-2
Recommended Storage : Store at Room Temperature.