missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Pyroglutamic Acid 97.0+%, TCI America™
Supplier: TCI America P0573500G
Specifications
L-Pyroglutamic Acid | |
160°C | |
C5H7NO3 | |
500 g | |
ODHCTXKNWHHXJC-UHFFFAOYNA-N | |
5-oxopyrrolidine-2-carboxylic acid | |
7405 | |
129.12 | |
Crystalline Powder |
98-79-3 | |
White | |
MFCD00005272 | |
l-pyroglutamic acid, pyroglutamic acid, pidolic acid, 5-oxo-l-proline, h-pyr-oh, 5-oxoproline, s-5-oxopyrrolidine-2-carboxylic acid, 2s-5-oxopyrrolidine-2-carboxylic acid, l-5-oxoproline, l-proline, 5-oxo | |
OC(=O)C1CCC(=O)N1 | |
129.12 | |
CHEBI:18183 | |
≥97.0% (T) |
Chemical Identifiers
98-79-3 | |
129.12 | |
ODHCTXKNWHHXJC-UHFFFAOYNA-N | |
7405 | |
5-oxopyrrolidine-2-carboxylic acid |
C5H7NO3 | |
MFCD00005272 | |
l-pyroglutamic acid, pyroglutamic acid, pidolic acid, 5-oxo-l-proline, h-pyr-oh, 5-oxoproline, s-5-oxopyrrolidine-2-carboxylic acid, 2s-5-oxopyrrolidine-2-carboxylic acid, l-5-oxoproline, l-proline, 5-oxo | |
CHEBI:18183 | |
OC(=O)C1CCC(=O)N1 |
Safety and Handling
EINECSNumber : (5)-0115
RTECSNumber : TW3710000
TSCA : Yes