3M™ L-Series Inner Shroud, L-226-2
Used in conjunction with 3M™ Outer Shroud, L-223, 3M™ Outer Shroud, L-225-2, 3M™ Outer Shroud, L-227, or 3M™ Outer Shroud, L-228-2.
Supplier: 3M™ 70071092806
Description
- The 3M™ L-Series Inner Shroud is used with a choice of outer shrouds making it compatible with the 3M™ Helmet, L-901 and the 3M™ Helmet, L-905 with proper respiratory protection.
Specifications
Headgear | |
Helmet with Faceshield |