missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals L-Serine, 99%
$78.25 - $629.47
Chemical Identifiers
CAS | 56-45-1 |
---|---|
Molecular Formula | C3H7NO3 |
Molecular Weight (g/mol) | 105.093 |
InChI Key | MTCFGRXMJLQNBG-REOHCLBHSA-N |
Synonym | l-serine, serine, s-2-amino-3-hydroxypropanoic acid, s-serine, beta-hydroxyalanine, h-ser-oh, l-ser, l---serine, serine, l, 2s-2-amino-3-hydroxypropanoic acid |
PubChem CID | 5951 |
ChEBI | CHEBI:17115 |
IUPAC Name | (2S)-2-amino-3-hydroxypropanoic acid |
SMILES | C(C(C(=O)O)N)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC132660250
|
Thermo Scientific Chemicals
132660250 |
25 g | Glass Bottle |
Each for $78.25
|
|
||||
AC132661000
|
Thermo Scientific Chemicals
132661000 |
100 g | Glass Bottle |
Each for $195.03
|
|
||||
AC132665000
|
Thermo Scientific Chemicals
132665000 |
500 g | Glass Bottle |
Each for $629.47
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
56-45-1 | |
105.093 | |
l-serine, serine, s-2-amino-3-hydroxypropanoic acid, s-serine, beta-hydroxyalanine, h-ser-oh, l-ser, l---serine, serine, l, 2s-2-amino-3-hydroxypropanoic acid | |
CHEBI:17115 | |
C(C(C(=O)O)N)O |
C3H7NO3 | |
MTCFGRXMJLQNBG-REOHCLBHSA-N | |
5951 | |
(2S)-2-amino-3-hydroxypropanoic acid |
Specifications
222.0°C | |
56-45-1 | |
5.0 to 6.5 (2.5% soln. at 25°C) | |
200ppm max. | |
Authentic | |
0.2% (105°C, 3 hrs) max. | |
0.5% (TLC) max. | |
Glass Bottle | |
15.2 | |
95% (c=10, H2O, 430nm) 1cm cellmin. | |
l-serine, serine, s-2-amino-3-hydroxypropanoic acid, s-serine, beta-hydroxyalanine, h-ser-oh, l-ser, l---serine, serine, l, 2s-2-amino-3-hydroxypropanoic acid | |
0150.0°C | |
1.6000g/mL | |
(2S)-2-amino-3-hydroxypropanoic acid | |
105.093 | |
5951 | |
105.09 | |
1.6 | |
L-Serine |
200ppm max. | |
1ppm max. | |
White | |
10ppm max. | |
10ppm max. | |
99% | |
C3H7NO3 | |
HOCH2CH(NH2)CO2H | |
200ppm max. | |
0.1% max. | |
Solubility in water: 250g/L (20°C). Other solubilities: insoluble in alcohol, ether and benzene, soluble in mineral acids, insoluble in organic solvents, insoluble in ethanol and diethylether | |
MTCFGRXMJLQNBG-REOHCLBHSA-N | |
C(C(C(=O)O)N)O | |
15.20 (20.00°C c=10,2N HCl) | |
25 g | |
CHEBI:17115 | |
99% | |
Crystalline Powder, Crystals or Crystalline Needles |
RUO – Research Use Only