missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Serine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1235522
Description
L-Serine methyl ester hydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields. It is also a used as a raw material in peptide synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsL-Serine methyl ester hydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields. It is also a used as a raw material in peptide synthesis.
Solubility
Soluble in water (50 mg/ml), and methanol
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture.
Specifications
155°C (decomposition) | |
5680-80-8 | |
MFCD00063680 | |
l-serine methyl ester hydrochloride, h-ser-ome.hcl, methyl l-serinate hydrochloride, s-methyl 2-amino-3-hydroxypropanoate hydrochloride, methyl 2s-2-amino-3-hydroxypropanoate hydrochloride, l-serine, methyl ester, hydrochloride, l-methyl serinate hcl, serine methyl ester hydrochloride, l-serine methyl ester hcl, h-ser-ome-hcl | |
NDBQJIBNNUJNHA-DFWYDOINSA-N | |
methyl (2S)-2-amino-3-hydroxypropanoate;hydrochloride | |
2723730 | |
98% | |
+5° (c=2 in methanol) |
L-Serine methyl ester hydrochloride | |
C4H10ClNO3 | |
3559591 | |
Soluble in water (50mg/ml),and methanol | |
COC(=O)C(CO)N.Cl | |
155.578 | |
155.58 | |
100 g | |
Hygroscopic |
Chemical Identifiers
5680-80-8 | |
155.578 | |
NDBQJIBNNUJNHA-DFWYDOINSA-N | |
2723730 | |
COC(=O)C(CO)N.Cl |
C4H10ClNO3 | |
MFCD00063680 | |
l-serine methyl ester hydrochloride, h-ser-ome.hcl, methyl l-serinate hydrochloride, s-methyl 2-amino-3-hydroxypropanoate hydrochloride, methyl 2s-2-amino-3-hydroxypropanoate hydrochloride, l-serine, methyl ester, hydrochloride, l-methyl serinate hcl, serine methyl ester hydrochloride, l-serine methyl ester hcl, h-ser-ome-hcl | |
methyl (2S)-2-amino-3-hydroxypropanoate;hydrochloride |
Safety and Handling
EINECSNumber : 227-140-7
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only