missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-(+)-Tartaric Acid (Granular/Certified ACS), Fisher Chemical™
Supplier: Thermo Fisher Scientific FLA314500
Specifications
L-(+)-Tartaric Acid | |
168°C | |
White | |
C4H6O6 | |
MFCD00064207 | |
FEWJPZIEWOKRBE-UHFFFAOYNA-N | |
2,3-dihydroxybutanedioic acid | |
444305 | |
150.09 | |
Certified ACS | |
500 g | |
Poly Bottle | |
<0.1 mbar at 20°C |
1.6 | |
87-69-4 | |
≥99.0 % | |
HOOC(CHOH)2COOH | |
l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid | |
OC(C(O)C(O)=O)C(O)=O | |
150.09 | |
CHEBI:15671 | |
≥99.0% | |
1.76g/mL | |
0.02% max. | |
Solid |
Chemical Identifiers
87-69-4 | |
150.09 | |
FEWJPZIEWOKRBE-UHFFFAOYNA-N | |
444305 | |
2,3-dihydroxybutanedioic acid |
C4H6O6 | |
MFCD00064207 | |
l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid | |
CHEBI:15671 | |
OC(C(O)C(O)=O)C(O)=O |