missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-tert-Leucine, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1370714

 View more versions of this product

Catalog No. AAL1370714


Explore available promotions

Description

Description

L-tert-Leucine is an essential amino acid and makes up one third of our muscle protein. It can be used as a food additive. It is used in the formation of sterols. It is used in the production of cobalt oxazoline palladacycles complex, which acts as a catalyst in the rearrangement of prochiral N-aryl trifluoroacetimidates to allylic amides yielded in significantly higher enantioselectivities with the (S)-(pR)-diasteroisomer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-tert-Leucine is an essential amino acid and makes up one third of our muscle protein. It can be used as a food additive. It is used in the formation of sterols. It is used in the production of cobalt oxazoline palladacycles complex, which acts as a catalyst in the rearrangement of prochiral N-aryl trifluoroacetimidates to allylic amides yielded in significantly higher enantioselectivities with the (S)-(pR)-diasteroisomer.

Solubility
Soluble in water and hydrchloric acid.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

>300°C
20859-02-3
MFCD00064218
l-tert-leucine, 2s-2-amino-3,3-dimethylbutanoic acid, 3-methyl-l-valine, l-valine,3-methyl, h-tle-oh, s-2-amino-3,3-dimethylbutanoic acid, 3-methyl-l-valin, l-2-amino-3,3-dimethylbutanoic acid, s-2-amino-3,3-dimethylbutyric acid
NPDBDJFLKKQMCM-SGAVLPGINA-N
(2S)-2-amino-3,3-dimethylbutanoic acid
131.18
+7° (c=5 in 5N HCl)
99%
L-tert-Leucine
C6H13NO2
1721824
Soluble in water and hydrchloric acid.
CC(C)(C)[C@H](N)C(O)=O
25 g
164608
131.18

Chemical Identifiers

20859-02-3
131.18
NPDBDJFLKKQMCM-SGAVLPGINA-N
164608
CC(C)(C)[C@H](N)C(O)=O
C6H13NO2
MFCD00064218
l-tert-leucine, 2s-2-amino-3,3-dimethylbutanoic acid, 3-methyl-l-valine, l-valine,3-methyl, h-tle-oh, s-2-amino-3,3-dimethylbutanoic acid, 3-methyl-l-valin, l-2-amino-3,3-dimethylbutanoic acid, s-2-amino-3,3-dimethylbutyric acid
(2S)-2-amino-3,3-dimethylbutanoic acid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 424-750-7

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only