missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-tert-Leucine Methyl Ester 98.0+%, TCI America™
$205.05 - $687.84
Chemical Identifiers
| CAS | 63038-26-6 |
|---|---|
| Molecular Formula | C7H15NO2 |
| Molecular Weight (g/mol) | 145.202 |
| MDL Number | MFCD00137409 |
| InChI Key | WCYLIGGIKNKWQX-RXMQYKEDSA-N |
| Synonym | H-Tle-OMe, Methyl (S)-2-Amino-3,3-dimethylbutyrate, (S)-2-Amino-3,3-dimethylbutyric Acid Methyl Ester |
| PubChem CID | 11309583 |
| IUPAC Name | methyl (2S)-2-amino-3,3-dimethylbutanoate |
| SMILES | CC(C)(C)C(C(=O)OC)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
L0255200MG
|
TCI America
L0255200MG |
200 mg |
Each for $205.05
|
|
|||||
|
L02551G
|
TCI America
L02551G |
1 g |
Each for $687.84
|
|
|||||
Chemical Identifiers
| 63038-26-6 | |
| 145.202 | |
| WCYLIGGIKNKWQX-RXMQYKEDSA-N | |
| 11309583 | |
| CC(C)(C)C(C(=O)OC)N |
| C7H15NO2 | |
| MFCD00137409 | |
| H-Tle-OMe, Methyl (S)-2-Amino-3,3-dimethylbutyrate, (S)-2-Amino-3,3-dimethylbutyric Acid Methyl Ester | |
| methyl (2S)-2-amino-3,3-dimethylbutanoate |
Specifications
| 63038-26-6 | |
| 69°C | |
| MFCD00137409 | |
| 1993 | |
| WCYLIGGIKNKWQX-RXMQYKEDSA-N | |
| methyl (2S)-2-amino-3,3-dimethylbutanoate | |
| 11309583 | |
| ≥98.0% (GC,T) | |
| L-tert-Leucine Methyl Ester |
| Colorless | |
| C7H15NO2 | |
| 200 mg | |
| H-Tle-OMe, Methyl (S)-2-Amino-3,3-dimethylbutyrate, (S)-2-Amino-3,3-dimethylbutyric Acid Methyl Ester | |
| CC(C)(C)C(C(=O)OC)N | |
| 145.202 | |
| 145.20 | |
| Liquid |
Safety and Handling
TSCA : No