missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Threonine Allo-free base, 990%, MP Biomedicals™

L-threonine is classified as an essential amino acid. Synthesized from homoserine phosphate by threonine synthase or from acetaldehyde and glycine by serine hydroxymethyltransferase

Supplier:  MP Biomedicals Inc 0210305380

 View more versions of this product

Catalog No. ICN10305380


Explore available promotions

Description

Description

Specifications

Specifications

L-Threonine
72-19-5
99%
MFCD00064270
AYFVYJQAPQTCCC-UHFFFAOYNA-N
2-amino-3-hydroxybutanoic acid
119.12
CHEBI:16857
99.5%
Allo Free
White
C4H9NO3
l-threonine, threonine, threonin, h-thr-oh, 2s,3r-2-amino-3-hydroxybutanoic acid, s-threonine, l---threonine, 2-amino-3-hydroxybutyric acid, threonine van, threoninum latin
CC(O)C(N)C(O)=O
100 g
6288
119.1
Crystalline Powder

Chemical Identifiers

72-19-5
119.12
AYFVYJQAPQTCCC-UHFFFAOYNA-N
6288
2-amino-3-hydroxybutanoic acid
C4H9NO3
MFCD00064270
l-threonine, threonine, threonin, h-thr-oh, 2s,3r-2-amino-3-hydroxybutanoic acid, s-threonine, l---threonine, 2-amino-3-hydroxybutyric acid, threonine van, threoninum latin
CHEBI:16857
CC(O)C(N)C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-774-1

Recommended Storage : Store at Room temperature (15 to 30°C).

SDS
Documents

Documents

Promotions

Promotions