missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Threonine (White Crystals or Crystalline Powder), Fisher BioReagents

Supplier:  Fisher BioReagents BP394100

Catalog No. BP394-100



Explore available promotions

Description

Description

This amino acid is suitable for use in tissue culture systems requiring additives.
Specifications

Specifications

255°C
0.02% max.
5.0 to 6.5
10ppm max.
Pass Test
99.0 to 101.0 %
Glass Bottle
15, 9534
l-threonine, threonine, threonin, h-thr-oh, 2s,3r-2-amino-3-hydroxybutanoic acid, s-threonine, l---threonine, 2-amino-3-hydroxybutyric acid, threonine van, threoninum latin
AYFVYJQAPQTCCC-UHFFFAOYNA-N
(2S,3R)-2-amino-3-hydroxybutanoic acid
100 g
CHEBI:16857
99.0 to 101.0%
L-Threonine
72-19-5
White
0.1% max.
0.2% max.
C4H9NO3
-27.6 to -29.0° (+ or -)
98.0% min.
Threonine
CC(O)C(N)C(O)=O
119.12
6288
119.12
Powder

Chemical Identifiers

72-19-5
119.12
l-threonine, threonine, threonin, h-thr-oh, 2s,3r-2-amino-3-hydroxybutanoic acid, s-threonine, l---threonine, 2-amino-3-hydroxybutyric acid, threonine van, threoninum latin
CHEBI:16857
CC(O)C(N)C(O)=O
C4H9NO3
AYFVYJQAPQTCCC-UHFFFAOYNA-N
6288
(2S,3R)-2-amino-3-hydroxybutanoic acid
Videos
Safety and Handling

Safety and Handling

Recommended Storage : RT

SDS Documents

Documents

Promotions

Promotions