missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ L-Tyrosine, Free Base, Calbiochem™,
Supplier: MilliporeSigma™ 6570100GM
Specifications
60-18-4 | |
C9H11NO3 | |
MFCD00002606 | |
OUYCCCASQSFEME-SVGMAFHSNA-N | |
(2S)-2-amino-3-(4-hydroxyphenyl)propanoic acid | |
181.19 | |
CHEBI:17895 | |
Solid |
White | |
Glass Bottle | |
l-tyrosine, tyrosine, s-tyrosine, p-tyrosine, h-tyr-oh, l-p-tyrosine, 2s-2-amino-3-4-hydroxyphenyl propanoic acid, 4-hydroxy-l-phenylalanine, l---tyrosine, tyrosine, l | |
N[C@@H](CC1=CC=C(O)C=C1)C(O)=O | |
100 g | |
6057 | |
≥98% (Titration) |