missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Tyrosine methyl ester hydrochloride, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2268322
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
187°C (decomposition) | |
3417-91-2 | |
MFCD00012607 | |
l-tyrosine methyl ester hydrochloride, h-tyr-ome.hcl, methyl l-tyrosinate hydrochloride, h-tyr-ome hcl, methyl tyrosinate hydrochloride, unii-w42m0mi271, tyrosine methyl ester hydrochloride, methyl l-tyrosinate hcl, s-methyl 2-amino-3-4-hydroxyphenyl propanoate hydrochloride, s-2-amino-3-4-hydroxy-phenyl-propionic acid methyl ester hydrochloride | |
Cl.COC(=O)[C@@H](N)CC1=CC=C(O)C=C1 | |
231.68 | |
231.68 | |
100 g | |
Hygroscopic |
L-Tyrosine methyl ester hydrochloride | |
C10H14ClNO3 | |
3917353 | |
VXYFARNRGZWHTJ-MVRPGTNWNA-N | |
methyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate;hydrochloride | |
76956 | |
≥98% | |
−5.2° (c= 2.4 in Water) |
Chemical Identifiers
3417-91-2 | |
231.68 | |
VXYFARNRGZWHTJ-MVRPGTNWNA-N | |
76956 | |
Cl.COC(=O)[C@@H](N)CC1=CC=C(O)C=C1 |
C10H14ClNO3 | |
MFCD00012607 | |
l-tyrosine methyl ester hydrochloride, h-tyr-ome.hcl, methyl l-tyrosinate hydrochloride, h-tyr-ome hcl, methyl tyrosinate hydrochloride, unii-w42m0mi271, tyrosine methyl ester hydrochloride, methyl l-tyrosinate hcl, s-methyl 2-amino-3-4-hydroxyphenyl propanoate hydrochloride, s-2-amino-3-4-hydroxy-phenyl-propionic acid methyl ester hydrochloride | |
methyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate;hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 222-313-3
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only