missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Valine, Multi-Compendial, U.S.P., J.T. Baker™
$2232.00 - $13520.11
Chemical Identifiers
CAS | 72-18-4 |
---|---|
Molecular Formula | C5H11NO2 |
Molecular Weight (g/mol) | 117.15 |
MDL Number | MFCD00064220 |
InChI Key | KZSNJWFQEVHDMF-BYPYZUCNSA-N |
Synonym | l-valine, valine, s-valine, s-2-amino-3-methylbutanoic acid, h-val-oh, 2-amino-3-methylbutyric acid, 2s-2-amino-3-methylbutanoic acid, s-2-amino-3-methylbutyric acid, l-alpha-amino-beta-methylbutyric acid, valinum latin |
PubChem CID | 6287 |
ChEBI | CHEBI:16414 |
IUPAC Name | (2S)-2-amino-3-methylbutanoic acid |
SMILES | CC(C)[C@H](N)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
02003244
|
Avantor J.T.Baker
209505 |
100 g | Poly Bottle |
Each for $2,232.00
|
|
||||
02003246
|
Avantor J.T.Baker
209506 |
1 kg | Poly Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
72-18-4 | |
117.15 | |
KZSNJWFQEVHDMF-BYPYZUCNSA-N | |
6287 | |
(2S)-2-amino-3-methylbutanoic acid |
C5H11NO2 | |
MFCD00064220 | |
l-valine, valine, s-valine, s-2-amino-3-methylbutanoic acid, h-val-oh, 2-amino-3-methylbutyric acid, 2s-2-amino-3-methylbutanoic acid, s-2-amino-3-methylbutyric acid, l-alpha-amino-beta-methylbutyric acid, valinum latin | |
CHEBI:16414 | |
CC(C)[C@H](N)C(O)=O |
Specifications
72-18-4 | |
MFCD00064220 | |
l-valine, valine, s-valine, s-2-amino-3-methylbutanoic acid, h-val-oh, 2-amino-3-methylbutyric acid, 2s-2-amino-3-methylbutanoic acid, s-2-amino-3-methylbutyric acid, l-alpha-amino-beta-methylbutyric acid, valinum latin | |
CC(C)[C@H](N)C(O)=O | |
117.15 | |
CHEBI:16414 | |
Multi-Compendial/USP | |
1L = 1.23kg |
C5H11NO2 | |
Poly Bottle | |
KZSNJWFQEVHDMF-BYPYZUCNSA-N | |
(2S)-2-amino-3-methylbutanoic acid | |
6287 | |
117.15 | |
100 g | |
L-Valine |