Fisherbrand™ Lab Grade Cylindrical Dewar Flask with Aluminum Housing and Handle
Borosilicate glass with silvered double walls and a convenient handle
Supplier: Fisherbrand™ FB1019610
Description
- Cylindrical- Wide Mouth
- Borosilicate Glass
- Silvered Double Walls
- PE Stopper Included
- For Use with Liquified Gases
- Aluminum Housing
- Lab Grade
- Coated Metal Handle
Specifications
Handle | |
5.63 in. | |
376 mm | |
Silver | |
4300 mL |
143 mm | |
Liquid Nitrogen | |
305 mm | |
Aluminum |