Fisherbrand™ Lab Grade Cylindrical Dewar Flask with Aluminum Housing and Handle
Borosilicate glass with silvered double walls and a convenient handle
Supplier: Fisherbrand™ FB1019626
Description
- Cylindrical- Wide Mouth
- Borosilicate Glass
- Silvered Double Walls
- PE Stopper Included
- For Use with Liquified Gases
- Aluminum Housing
- Lab Grade
- Coated Metal Handle
Specifications
Handle | |
2.75 in. | |
195 mm | |
Aluminum |
70 mm | |
Liquid Nitrogen | |
125 mm | |
350 mL |