Fisherbrand™ Lab Grade Cylindrical Dewar Flask with Aluminum Housing and Handle
Borosilicate glass with silvered double walls and a convenient handle
Supplier:  Fisherbrand™ FB101969
 View more versions of this product
Catalog No. FB101969
Explore available promotions
Description
Description
  • Cylindrical- Wide Mouth
  • Borosilicate Glass
  • Silvered Double Walls
  • PE Stopper Included
  • For Use with Liquified Gases
  • Aluminum Housing
  • Lab Grade
  • Coated Metal Handle
Specifications
Specifications
Handle
4.75 in.
256 mm
Silver
1900 mL
120 mm
Liquid Nitrogen
190 mm
Aluminum