TrippNT Lab Organizers DFS Item
TrippNT Lab Organizers, 30-Column Cabinet, For Use With: Protects columns and helps maintain inventory
Supplier:  TrippNT TRP50664
 View more versions of this product
Catalog No. 50856152
Explore available promotions
Description
Description
TrippNT Lab Organizers, 30-Column Cabinet, For Use With: Protects columns and helps maintain inventory
Lab organizers are available in various styles and sizes; Click on individual items for description and specifications
Specifications
Specifications
HPLC Personal Supplies Organizer
Provides storage for 10 HPLC columns, built in tool holder, book holder and lab wiper dispenser, space for your logbook or calibration manual