missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eagle™ Lab Pack Open Head Poly Drum with 2 in. Bung Holes, 55 gal.
Drum is constructed of blow-molded high-density polyethylene (HDPE) with UV inhibitors, is durable and lightweight, as well as weather and chemical resistant. Bung lid includes two, 2 in. holes and is ideal for safe storage, as well as handling or transportation of hazardous materials.
$188.97 - $198.06
Specifications
Product Type | Drum |
---|---|
Closure Included | Yes |
Capacity (English) | 55 gal. |
Capacity (Metric) | 208.48 L |
For Use With (Application) | Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration |
Catalog Number | Mfr. No. | Closure Type | Color | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Closure Type | Color | Price | Quantity | ||||
19338480
|
Eagle™
1656MBKBRBG2 |
Metal Bolt Ring | Black |
Each for $192.42
|
|
||||
19338479
|
Eagle™
1656MBBRBG2 |
Metal Bolt Ring | Blue |
Each for $198.06
|
|
||||
19338478
|
Eagle™
1656MBRBG2 |
Metal Bolt Ring | Yellow |
Each for $192.42
|
|
||||
19338477
|
Eagle™
1656MBKBG2 |
Metal Lever-lock | Black |
Each for $188.97
|
|
||||
19338476
|
Eagle™
1656MBBG2 |
Metal Lever-lock | Blue |
Each for $194.41
|
|
||||
19338475
|
Eagle™
1656MBG2 |
Metal Lever-lock | Yelllow |
Each for $190.39
|
|
||||
Description
- Constructed from blow-molded high-density polyethylene (HDPE) with UV inhibitors
- For use with smaller non-damaged bottles, cans, carboys and 5 gal. pails of non-leaking hazardous materials
- Bung lid includes two, 2 in. holes
- If used with leaking materials, drums must be packed with sufficient sorbents
- No free liquids
Specifications
Drum | |
55 gal. | |
Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration | |
Open | |
91 cm | |
23.5 lb. |
Yes | |
208.48 L | |
HDPE | |
35.8 in. | |
2 in. (Bung Holes) |