missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eagle™ Lab Pack Open Head Poly Drum, 55 gal.

Drum is constructed of blow-molded high-density polyethylene (HDPE) with UV inhibitors, is durable and lightweight, as well as weather and chemical resistant. Ideal for safe storage, handling or transportation of hazardous materials, spill containment, and bio waste disposal.

Supplier:  Eagle™ 1656M

 View more versions of this product

Catalog No. 19338468


Explore available promotions

Description

Description

  • Constructed from blow-molded high-density polyethylene (HDPE) with UV inhibitors
  • For use with smaller non-damaged bottles, cans, carboys and 5 gal. pails of non-leaking hazardous materials
  • If used with leaking materials, drums must be packed with sufficient sorbents
  • No free liquids
Specifications

Specifications

Drum
Metal Lever-lock
208.48 L
Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration
Open
91 cm
23.5 lb.
Yes
55 gal.
Yellow
HDPE
35.8 in.
21 in. (Top), 22.5 in. (Bottom)
SDS
Documents

Documents

Promotions

Promotions