missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eagle™ Lab Pack Open Head Poly Drum, 55 gal.
Drum is constructed of blow-molded high-density polyethylene (HDPE) with UV inhibitors, is durable and lightweight, as well as weather and chemical resistant. Ideal for safe storage, handling or transportation of hazardous materials, spill containment, and bio waste disposal.
$194.06 - $227.50
Specifications
Product Type | Drum |
---|---|
Closure Included | Yes |
Capacity (English) | 55 gal. |
Capacity (Metric) | 208.45 L |
For Use With (Application) | Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration |
Catalog Number | Mfr. No. | Closure Type | Color | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Closure Type | Color | Price | Quantity | ||||
19338403
|
Eagle™
1655B |
Plastic Lever-lock | Blue |
Each for $194.06
|
|
||||
19338402
|
Eagle™
1655 |
Plastic Lever-lock | Yellow |
Each for $201.82
|
|
||||
19338412
|
Eagle™
1655MBLK |
Metal Lever-lock | Black |
Each for $209.63
|
|
||||
19338406
|
Eagle™
1655MB |
Metal Lever-lock | Blue |
Each for $210.83
|
|
||||
19338405
|
Eagle™
1655M |
Metal Lever-lock | Yellow |
Each for $211.85
|
|
||||
19338414
|
Eagle™
1655MBLKBR |
Metal Bolt Ring | Black |
Each for $210.53
|
|
||||
19338408
|
Eagle™
1655MBBR |
Metal Bolt Ring | Blue |
Each for $227.50
|
|
||||
Description
- Constructed from blow-molded high-density polyethylene (HDPE) with UV inhibitors
- For use with smaller non-damaged bottles, cans, carboys and 5 gal. pails of non-leaking hazardous materials
- If used with leaking materials, drums must be packed with sufficient sorbents
- No free liquids
Specifications
Drum | |
55 gal. | |
Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration | |
Open | |
98 cm | |
23.5 lb. |
Yes | |
208.45 L | |
HDPE | |
38.6 in. | |
23.75 in. (Top), 19.4 in. (Bottom) |